Drukuj
Znaczniki do drukarek etykiet BBP3x/i3300, typ B-7643
Dostawca: Brady
195554 195555 195556 195557 195558 195559 195560 195561
BRDY195554EA 1370 PLN
BRDY195554 BRDY195555 BRDY195556 BRDY195557 BRDY195558 BRDY195559 BRDY195560 BRDY195561
Znaczniki do drukarek etykiet BBP3x/i3300, typ B-7643
Etykiety
Thermoplastic polyether polyurethane is a zero-halogen high-density, thermoplastic cable marker designed for dot matrix or thermal-transfer printing.
  • Meets the requirements of a halogen-free material per IEC 61249-2-21 (2003-11)
  • Excellent chemical, oil and water resistance
  • RoHS compliant using EU directive 2002/95/EC
  • Operating temperature of −40 to +90 °C (−40 to +193 °F)