Drukuj
Etykiety do drukarek etykiet BBP33/i3300, typ B-7598
Dostawca: Brady
197539 197540 197541 197542 197543 197544 197545 197546 197547 197548 197549 197550 197551 197552 197553 197554 197555 197556 197558 197559 197560 197561 197562 197563 197565
BRDY197539EA 3590 PLN
BRDY197539 BRDY197540 BRDY197541 BRDY197542 BRDY197543 BRDY197544 BRDY197545 BRDY197546 BRDY197547 BRDY197548 BRDY197549 BRDY197550 BRDY197551 BRDY197552 BRDY197553 BRDY197554 BRDY197555 BRDY197556 BRDY197558 BRDY197559 BRDY197560 BRDY197561 BRDY197562 BRDY197563 BRDY197565
Etykiety do drukarek etykiet BBP33/i3300, typ B-7598
Etykiety
These B-7598 thermal transfer printable polyester tag offers a transfer adhesive it can be used for patch panel identification and equipment identification.
  • No added halogens
  • Made of polyester material
Ideal for marking of electrical cable and wires of various diameters.
Certyfikaty: RoHS compliance with Directive 2011/65/EU and 2015/863. Part does not contain an SVHC. Parts supplied in white, black or silver are Halogen Free as per IEC 61249-2-21 (2003-11).
Sposób pakowania: 1000 tags per roll.