Drukuj
Etykiety do drukarek termotransferowych BBP33/i3300, typ B-7643
Dostawca: Brady
BRDY197499EA 2010 PLN
BRDY197499 BRDY197498 BRDY197497 BRDY197496 BRDY197495 BRDY197494 BRDY197493 BRDY197492 BRDY197491 BRDY197490 BRDY197504 BRDY197503 BRDY197501 BRDY197489 BRDY197500 BRDY197488 BRDY197487 BRDY197486 BRDY197485 BRDY197484 BRDY197483 BRDY197482 BRDY197481 BRDY197509 BRDY197508 BRDY197507 BRDY197506 BRDY197505
Etykiety do drukarek termotransferowych BBP33/i3300, typ B-7643
Etykiety
These B-7643 cable tags offers excellent fire safety characteristics combined with minimal smoke emission.
  • No added halogens
  • Made of thermoplastic polyether polyurethane material
Ideal for data and telecommunications labelling, wire and cable labelling.
Certyfikaty: RoHS compliance with Directive 2011/65/EU and 2015/863. Halogen free per IEC 61249-2-21 (2003-11), Adherence: SAE-AS81531-1998, Flame retardant: MIL-STD-202G.