Drukuj
Rękawice dla przemysłu spożywczego, HyFlex® 74-500
Dostawca: Ansell
74-500/9 74-500/10 74-500/7 74-500/8 74-500/11 74-500/6
ANSE74-500/9EA 1230 PLN
ANSE74-500/9 ANSE74-500/10 ANSE74-500/7 ANSE74-500/8 ANSE74-500/11 ANSE74-500/6
Rękawice dla przemysłu spożywczego, HyFlex® 74-500
Rękawice
INTERCEPT Technology delivers highest cut protection level F throughout entire glove and cuff; even after repeat laundering.
  • Constructed of fiberglass, stainless steel, HPPE, polyester & polyamide
  • Ambidexterous
  • Extended cuff expanding cut protection to wrists
  • High durability design retains dimensions and protection even after repeat laundering
  • Pre-washed; maintains protection, comfort and fit even after repeat washes
Recommended for: Dairy processing, fruit and vegetable processing, poultry processing
Certyfikaty: EN 388:2016 (4X4XF); Cat. II
Food contact approved
Sposób pakowania: 12 pieces per polybag.