Drukuj
Rękawice odporne na zimno, Showa TEMRES® 282
Dostawca: Showa
282-9/L 282-8/M 282-10/XL 282-11/XXL
112-5758EA 1930 PLN
112-5758 112-5757 112-5755 112-5756
Rękawice odporne na zimno, Showa TEMRES® 282
Rękawice
These breathable, waterproof and insulated PU coated gloves provide warmth and protection from liquids in cold conditions.
  • Full double polyurethane (PU) coating
  • Breathable membrane fixed between coating and liner
  • Micro roughened coating finish
  • Anatomical shape and seamless knit for optimum comfort
  • High levels of handling precision and grip
  • Ergonomic design that replicates the natural curvature of a human hand and so reduces hand fatigue
Features fixed insulated acrylic liner which protects from cold ensuring warmth and comfort. Impermeable coating prevents ingress from water and liquids. Breathable technology allows warm air and moisture from inside to escape to keep hands dry.
Applications include agriculture, construction, cold storage, forestry and fishing, warehousing and distribution, wet work.
Certyfikaty: EN 388:2016 (4221X); EN 388:2003 (4221); EN 511:2006 (021); Cat. II
Sposób pakowania: 10 pairs per pack.
Uwaga: Do not use in cases of chemical, electrical or thermal risk. Store away from light and humidity.