Drukuj
Łaźnie wodne ogólnego przeznaczenia Precision
Dostawca: THERMO ELECTRON LED
TSGP02 TSGP05 TSGP10 TSGP20 TSGP28 TSGP15D
HAKATSGP02EA 4580 PLN
HAKATSGP02 HAKATSGP05 HAKATSGP10 HAKATSGP20 HAKATSGP28 HAKATSGP15D
Łaźnie wodne ogólnego przeznaczenia Precision
Łaźnie Łaźnie wodne bez mieszania
Precision™ general purpose water baths are designed to maintain water temperature from ambient to 100 °C.
  • Smaller footprint, compared to previous models, frees up valuable benchtop space
  • Advanced microprocessor controller is designed for extended functionality
  • Protects work with audible alarms
  • Conveniently save commonly used settings with four temperature presets
These rugged, high-performance baths range from 2 to 28 litres, including shallow models. Over-temperature safety circuitry is designed to prevent thermal runaway, while new auto-on and auto-off timers allow you to optimize operation schedules. Benefit from outstanding chemical and corrosion resistance with epoxy powder-coated exterior, easily clean the chamber with its seamless stainless steel interior.
The temperature uniformity lies at a range of ±0,2 °C.
Electrical information: 100 to 115 V/200 to 230 V, 50/60 Hz
Certyfikaty: cULus Listed, CE compliant, US FDA Class I Medical Device.
Dodatkowe informacje: Baths come with clear polycarbonate gable cover, diffuser tray, drain hose and rubber duck. Includes a setting for thermal beads. Comes with N5-15 Plug.