Drukuj
Szybkie testy kolorymetryczne, potas, VISOCOLOR® ECO
Dostawca: MACHEREY-NAGEL
931032 931232
MANA931032EA 604 PLN
MANA931032 MANA931232
Szybkie testy kolorymetryczne, potas, VISOCOLOR® ECO
Zestawy do testów środowiskowych
Determination of potassium by turbidity measurement.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
The natural potassium content in ground water is generally about 1 to 2 mg/l K+. Higher values may indicate faecal contaminations, but can also originate from potassium fertilisers. For the growth of plants and animals potassium is an essential factor. Especially in agriculture the determination of potassium therefore gains increasing importance.
Suitable for sea water testing after dilution.