Szybkie testy kolorymetryczne, chlorek, VISOCOLOR®
Dostawca: MACHEREY-NAGEL
915004
915204
931018
931218
MANA915004EA
371
PLN
MANA915004
MANA915204
MANA931018
MANA931218
Szybkie testy kolorymetryczne, chlorek, VISOCOLOR®
Zestawy do testów środowiskowych
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
- Portable
- Environmentally friendly
- Convenient handling
- Application-oriented measuring ranges
Chloride ions occur in all natural waters. Their concentration depends on the geological and local situation. In waste waters and polluted rivers the chloride concentration can reach high values.