Drukuj
Buty ochronne ESD, sznurowane, uvex 2 xenova®, 9507
Dostawca: Uvex
uvex 2 xenova®
UVEX9507.7/38EA 582 PLN
UVEX9507.7/38 UVEX9507.7/39 UVEX9507.8/46 UVEX9507.8/47 UVEX9507.9/39 UVEX9507.8/48 UVEX9507.8/49 UVEX9507.9/38 UVEX9507.7/52 UVEX9507.8/42 UVEX9507.8/43 UVEX9507.8/44 UVEX9507.8/45 UVEX9507.9/51 UVEX9507.9/50 UVEX9507.8/40 UVEX9507.7/50 UVEX9507.9/52 UVEX9507.7/51 UVEX9507.8/41 UVEX9507.7/49 UVEX9507.8/39 UVEX9507.9/48 UVEX9507.7/45 UVEX9507.7/46 UVEX9507.9/47 UVEX9507.7/47 UVEX9507.9/49 UVEX9507.8/38 UVEX9507.7/48 UVEX9507.9/44 UVEX9507.7/41 UVEX9507.9/43 UVEX9507.7/42 UVEX9507.9/46 UVEX9507.7/43 UVEX9507.7/44 UVEX9507.9/45 UVEX9507.9/40 UVEX9507.8/50 UVEX9507.9/42 UVEX9507.8/51 UVEX9507.7/40 UVEX9507.8/52 UVEX9507.9/41
Buty ochronne ESD, sznurowane, uvex 2 xenova®, 9507
Obuwie i buty
Metal-free, lightweight, flexible and robust safety shoe made from water-repellent, full-grain smooth leather upper material, soft padded tongue and collar, and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Anti-static comfort insole with moisture control system; orthopaedic insoles
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/red
Certyfikaty: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC