Drukuj
Buty ochronne ESD, sznurowane, Mason Pro Rubber Low
Dostawca: ELTEN
7282101/40 7282101/41 7282101/42 7282101/43 7282101/44 7282101/45 7282101/46 7282101/47 7282101/48 7282102/40 7282102/41 7282102/42 7282102/43 7282102/44 7282102/45 7282102/46 7282102/47 7282102/48 7282103/40 7282103/41 7282103/42 7282103/43 7282103/44 7282103/45 7282103/46 7282103/47 7282103/48
ELTN7282101/40EA 446 PLN
ELTN7282101/40 ELTN7282101/41 ELTN7282101/42 ELTN7282101/43 ELTN7282101/44 ELTN7282101/45 ELTN7282101/46 ELTN7282101/47 ELTN7282101/48 ELTN7282102/40 ELTN7282102/41 ELTN7282102/42 ELTN7282102/43 ELTN7282102/44 ELTN7282102/45 ELTN7282102/46 ELTN7282102/47 ELTN7282102/48 ELTN7282103/40 ELTN7282103/41 ELTN7282103/42 ELTN7282103/43 ELTN7282103/44 ELTN7282103/45 ELTN7282103/46 ELTN7282103/47 ELTN7282103/48
Buty ochronne ESD, sznurowane, Mason Pro Rubber Low
Obuwie i buty
ESD safety ankle shoes made from wear resistant breathable cowhide leather with fabric lining, reflective materials, a full padded bellows tongue and a dual density rubber/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • TPU scuff cap
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected, for low and high temperatures and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certyfikaty: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, HI/HRO/SRC, form A