Drukuj
VWR®, butelki laboratoryjne, wąska szyjka
Dostawca: VWR Collection
VWRI215-1592-EEA 200 PLN
VWRI215-1592-E VWRI215-1593-E VWRI215-1594-E VWRI215-1595-E VWRI215-1596-E
VWR®, butelki laboratoryjne, wąska szyjka
Butelki i butle Butelki szklane
Borosilicate 3.3 glass, without screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Economical alternative for a wide range of laboratory applications. Round, with GL 45 thread and white graduations.
Certyfikaty: ISO 4796
Informacje do zamówienia: PP screw cap and pouring ring must be ordered separately.