Drukuj
VISOCOLOR® ECO Colorimetric Test, Cyanide
Dostawca: MACHEREY-NAGEL
931022 931222
MANA931022EA 404 PLN
MANA931022 MANA931222
VISOCOLOR® ECO Colorimetric Test, Cyanide
Zestawy do testów środowiskowych
Test kit for the determination of cyanide in water samples.
  • Combines simple implementation with high measurement reliability
For visual determination in comparison to a colour chart or for photometric measurement with our compact photometers.
Sea water analysis: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Hazardous material: Yes