Drukuj
VWR®, Obiektywy do IOS W-Plan MET
Dostawca: VWR Collection
630-2159EA 2550 PLN
630-2159 630-2161 630-2162
VWR®, Obiektywy do IOS W-Plan MET
Obiektywy do mikroskopów
These objectives deliver precise performance without the need of the cover slide, being NCG (no cover glass). They are intended to be used in metallurgical and epi-illumination applications. Ideal for routine laboratory needs.
Designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.