Drukuj
VWR®, Obiektywy z kontrastem fazowym IOS W-Plan Phase Contrast
Dostawca: VWR Collection
630-1976EA 1150 PLN
630-1976 630-1977 630-1978 630-1979
VWR®, Obiektywy z kontrastem fazowym IOS W-Plan Phase Contrast
Obiektywy do mikroskopów
IOS W-Plan PH objectives deliver outstanding performance in phase contrast technique, providing a great contrast generally required in high-level education and routine laboratory needs.
The objectives are designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.