Drukuj
Cyrkulatory chłodzące/grzewcze, seria, DYNEO™ DD
Dostawca: JULABO GmbH
9021704.D 9021701 9021702 9021703 9021704 9021705 9021707 9021701.A 9021702.A 9021703.A 9021708 9021704.A 9021705.A 9021707.A 9021708.A 9021701.D 9021702.D 9021703.D 9021705.D 9021707.D 9021708.D 9021713.N1 9021713.N1.A 9021713.N1.D 9021713.S1 9021713.S1.A 9021713.S1.D 9021714.N1 9021714.N1.A 9021714.N1.D 9021714.S1 9021714.S1.A 9021714.S1.D 9021716.N1 9021716.N1.A 9021716.N1.D 9021715.N1 9021715.N1.A 9021715.N1.D 9021717.N1 9021717.N1.A 9021717.N1.D 9021701.N1 9021701.N1.A 9021701.N1.D 9021728.N1 9021728.N1.A 9021728.N1.D
461-1418EA 36220 PLN
461-1418 461-1388 461-1389 461-1390 461-1391 461-1392 461-1394 461-1401 461-1402 461-1403 461-1395 461-1404 461-1405 461-1407 461-1408 461-1415 461-1416 461-1417 461-1419 461-1421 461-1422 JULA9021713.N1 JULA9021713.N1.A JULA9021713.N1.D JULA9021713.S1 JULA9021713.S1.A JULA9021713.S1.D JULA9021714.N1 JULA9021714.N1.A JULA9021714.N1.D JULA9021714.S1 JULA9021714.S1.A JULA9021714.S1.D JULA9021716.N1 JULA9021716.N1.A JULA9021716.N1.D JULA9021715.N1 JULA9021715.N1.A JULA9021715.N1.D JULA9021717.N1 JULA9021717.N1.A JULA9021717.N1.D JULA9021701.N1 JULA9021701.N1.A JULA9021701.N1.D JULA9021728.N1 JULA9021728.N1.A JULA9021728.N1.D
Cyrkulatory chłodzące/grzewcze, seria, DYNEO™ DD
Termostaty cyrkulacyjne Systemy cyrkulacyjne, otwarte Cyrkulatory z grzaniem/chłodzeniem
DYNEO™ DD refrigerated circulators are powerful cooling units suitable for internal or external applications between –50 and +200 °C. The circulators function precisely and reliably even at elevated ambient temperatures up to +40 °C. The multilingual 8,89 cm colour display and unique rotary knob allow straightforward and intuitive operation. Units have ergonomic handles and a built in drain tap for easy and safe drainage.
  • Optimised cooling coil design saves space in the bath tank
  • Powerful and infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Large colour TFT display, multilingual user interface and USB port
  • Central rotary knob (controller) simplifies operation
  • Integrated programmer for 8×60 program steps
  • Integrated external PT100 connection
  • RS232 interface or analogue interface (optional)
  • Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
DD: 8 to 27 L/min, 0,1 to 0,7 bar.
Certyfikaty: Classification III according to DIN 12876-1
IP protection class according to DIN EN 60529: IP 21.
Informacje do zamówienia: An extensive range of accessories mean that the circulators can be modularly and individually adapted to applications.
Dodatkowe informacje: Supplied with bath cover and two each barbed fitting for tubing 8 and 12 mm inner dia. (pump connections M16×1 male).
Uwaga: NOTE: Cooling capacity at +20, 0, −10, −20, −30 and −40 °C.