Drukuj
Płaszcze grzewcze, seria WHG2
Dostawca: MOHR
WH/G2/50RC WH/G2/100RC WH/G2/500RC WH/G2/1000RC WH/G2/2000RC WH/G2/3000RC WH/G2/4000RC WH/G2/5000RC WH/G2/6000RC
454-1106EA 0 PLN
454-1106 454-1110 454-1118 454-1122 454-1126 454-1130 454-1134 454-1138 454-1142
Płaszcze grzewcze, seria WHG2
Urządzenia grzewcze Czasze grzejne
These heating mantles feature crocheted outer jacket for round bottom flasks.
  • Hand crocheted heating mantles made of glass silk
  • With detachable plug-in connection of 0,5 m cable length
  • RCD safety plug of 1,0 m cable length
  • T max = 450 °C
The 25 and 50 ml heating mantles are with slide switch, 100 to 1000 ml with series/parallel slide switch and from 2000 ml with series/parallel rotary switch.