Drukuj
Kolba Synthware™ Kuderna-Danish
Dostawca: Merck
SIALSYNF984250-1EAEA 356 PLN
SIALSYNF984250-1EA
Kolba Synthware™ Kuderna-Danish
Koncentratory
Flask with top joint 24/40 outer while the lower joint is a 19/22 inner.