Drukuj
Kolumny kapilarne, OPTIMA® FFAPplus
Dostawca: MACHEREY-NAGEL
726241.30 726241.60 726242.30 726242.60 726243.30 726243.60 726246.30 726246.60
MANA726241.30EA 2840 PLN
MANA726241.30 MANA726241.60 MANA726242.30 MANA726242.60 MANA726243.30 MANA726243.60 MANA726246.30 MANA726246.60
Kolumny kapilarne, OPTIMA® FFAPplus
Kolumny chromatograficzne
Polar GC phase with optimised crosslinking for lower column bleeding.
  • Suitable for MS detectors
  • Recommended for FAME and free carboxylic acids
  • Maximum temperature for isothermal operation 250 °C
  • Maximum temperature for short isotherms in a temperature program 260 °C
Polyethylene glycol 2-nitroterephthalate phase.
USP code G35