Drukuj
Kolumny HPLC, analiza kwasów nukleinowych, NUCLEOGEN® DEAE
Dostawca: MACHEREY-NAGEL
736400.40 736596.40 736598 736601 736597.100 736599.100 736602.100
MANA736400.40EA 1030 PLN
MANA736400.40 MANA736596.40 MANA736598 MANA736601 MANA736597.100 MANA736599.100 MANA736602.100
Kolumny HPLC, analiza kwasów nukleinowych, NUCLEOGEN® DEAE
Kolumny chromatograficzne
Silica-based phase for HPLC separation of nucleic acids.
  • Robust spherical silica gel
  • DEAE anion exchanger
  • Separation of oligonucleotides, DNA and RNA fragments
NUCLEOGEN® 60-7 DEAE is recommended for the separation of oligonucleotides up to chain lengths of 40 bases.
NUCLEOGEN® 500-7 DEAE can be applied for the separation of tRNA, mRNA, and viroids in the intermediate molecular weight range (25000 to 1000000 daltons).
NUCLEOGEN® 4000-7 DEAE is suitable for the separation of plasmids, DNA restriction fragments, ribosomal RNA, messenger RNA and viral RNA (1 to 50 megadaltons).
CC guard columns require the CC column holder (30 mm).