Drukuj
Etykiety do drukarek termotransferowych, typ B-430, WorkHorse™
Dostawca: Brady
030216 030189 620362 030190 030188
BRDY030216EA 1560 PLN
BRDY030216 BRDY030189 BRDY620362 BRDY030190 BRDY030188
Etykiety do drukarek termotransferowych, typ B-430, WorkHorse™
Etykiety
Printable, clear polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.