Drukuj
Obuwie ochronne ESD, sznurowane, Maddox S1P
Dostawca: ELTEN
729571/36 729571/37 729571/38 729571/39 729571/40 729571/41 729571/42 729571/43 729571/44 729571/45 729571/46 729571/47 729571/48 729571/35 729551/40 729551/41 729551/42 729551/43 729551/44 729551/45 729551/46 729551/47 729551/48
ELTN729571/36EA 501 PLN
ELTN729571/36 ELTN729571/37 ELTN729571/38 ELTN729571/39 ELTN729571/40 ELTN729571/41 ELTN729571/42 ELTN729571/43 ELTN729571/44 ELTN729571/45 ELTN729571/46 ELTN729571/47 ELTN729571/48 ELTN729571/35 ELTN729551/40 ELTN729551/41 ELTN729551/42 ELTN729551/43 ELTN729551/44 ELTN729551/45 ELTN729551/46 ELTN729551/47 ELTN729551/48
Obuwie ochronne ESD, sznurowane, Maddox S1P
Obuwie i buty
Leather-free ESD safety shoes made from microfibre/textile material, a full padded bellows tongue plus a dual-density TPU/PU sole (Maximmo).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole SPORTIVE ESD
  • Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where there is a risk of penetration from pointed and sharp objects.
Colour: Grey/blue or blue
Certyfikaty: EN 61340-5-1 (ESD); EN ISO 20345; S1P, SRC.