Drukuj

Anti-CALB1 Chicken Polyclonal Antibody

Dostawca: Biosensis
C-1798-50
BSENC-1798-50EA 1570 PLN
BSENC-1798-50
Anti-CALB1 Chicken Polyclonal Antibody
Przeciwciała
Buffers cytosolic calcium. May stimulate a membrane Ca2+-ATPase and a 3',5'-cyclic nucleotide phosphodiesterase. Ref: uniprot.org. Antibody is specific for calbindin and does not recognize closely related proteins parvalbumin and calretinin as determined by Western Blotting.

Western blotting (1:1,000-1:5,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CALB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Cow
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR