Acetylokoenzym A
Dostawca: Merck
An essential cofactor in enzymatic acetyl transfer reactions.
- Moisture-sensitive
- Desiccated and stored at −20 °C
Acetyl coenzyme A is suitable in CAT reporter assay for the measurement of chloramphenicol acetyl transferase by incubating the cell extract with 0,1 μCi [14C] chloramphenicol and 0,5 mM acetyl-CoA. Acetyl CoA is soluble in deionised water at 100 mg/ml. Acetyl CoA is stable in neutral and moderately acidic solutions but will hydrolyse in alkaline and strongly acidic solutions.