Drukuj
Acetylokoenzym A
Dostawca: Merck
A2181-25MG A2181-100MG A2181-10MG A2181-1MG A2181-5MG
SIALA2181-25MGEA 2430 PLN
SIALA2181-25MG SIALA2181-100MG SIALA2181-10MG SIALA2181-1MG SIALA2181-5MG
Acetylokoenzym A
Enzymy
An essential cofactor in enzymatic acetyl transfer reactions.
  • Moisture-sensitive
  • Desiccated and stored at −20 °C
Acetyl coenzyme A is suitable in CAT reporter assay for the measurement of chloramphenicol acetyl transferase by incubating the cell extract with 0,1 μCi [14C] chloramphenicol and 0,5 mM acetyl-CoA. Acetyl CoA is soluble in deionised water at 100 mg/ml. Acetyl CoA is stable in neutral and moderately acidic solutions but will hydrolyse in alkaline and strongly acidic solutions.