Drukuj
Pełne maski ochronne na twarz, Advantage 3100
Dostawca: M S A
10027725 10027724 10027723
MSAA10027725EA 1360 PLN
MSAA10027725 MSAA10027724 MSAA10027723
Pełne maski ochronne na twarz, Advantage 3100
Maski oddechowe Maski wielokrotnego użytku Maski pełnotwarzowe
The Advantage 3100 full mask series provides high protection and unique comfort. The soft sealing line made from hypoallergenic silicone provides a pressure free fit.
  • Large, optically corrected lens ensures a clear, undistorted view
  • Grey-blue colour gives the mask an aesthetic appearance
  • Quick, simple donning without painful hair pulling
  • Patented head harness adapts perfectly to the facial contours
Suitable for use with all filter series with wire connection according to EN 148.
Certyfikaty: EN 136 class 3