Drukuj
Kolumny do chromatografii powinowactwa, HiTrap™ IXSelect
Dostawca: Cytiva
17-3714-11 17-3714-12
GEHE17-3714-11EA 5860 PLN
GEHE17-3714-11 GEHE17-3714-12
Kolumny do chromatografii powinowactwa, HiTrap™ IXSelect
Kolumny chromatograficzne
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
  • IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.