Drukuj
Obuwie ochronne do kostki, sznurowane, Macsole X, MX 300 GT/MX 400 GT, Heckel
Dostawca: UVEX
6393337 6393338 6393339 6393340 6393341 6393342 6393343 6393345 6393346 6393347 6393348 6395346 6395344 6395345 6395337 6395338 6395339 6395340 6395341 6395342 6395343 6395347 6395348
UVEX6393337EA 0 PLN
UVEX6393337 UVEX6393338 UVEX6393339 UVEX6393340 UVEX6393341 UVEX6393342 UVEX6393343 UVEX6393345 UVEX6393346 UVEX6393347 UVEX6393348 UVEX6395346 UVEX6395344 UVEX6395345 UVEX6395337 UVEX6395338 UVEX6395339 UVEX6395340 UVEX6395341 UVEX6395342 UVEX6395343 UVEX6395347 UVEX6395348
Obuwie ochronne do kostki, sznurowane, Macsole X, MX 300 GT/MX 400 GT, Heckel
Obuwie i buty
Sturdy, 100% waterproof safety ankle boots made from water resistant full grain leather, padded tongue and collar, closure system with metallic hooks and eyelets, reflective piping plus a double-density PU/rubber sole, resistant to oils, hydrocarbons, and chemicals also heat resistant up to 300 °C.
  • GORE-TEX breathable, waterproof membrane
  • Anatomically-shaped hygienic insole
  • Shock-absorbent heel
  • Self-cleaning profile with excellent grip
  • Injection-moulded PU scuff cap
  • Metal-free protective toe cap (200 J)
  • Metal-free puncture resistant midsole
Suitable for outdoor work in humid conditions, construction, transportation networks, energy, chemicals and forestry.
Colour: Black, brown
Weight : 715 g
Certyfikaty: EN ISO 20345: 2011; S3, CI, HI, WR, HRO, SRC