Drukuj
Obuwie ochronne ESD, sznurowane, Basic, jo_Colour Orange Low, Jori
Dostawca: ELTEN
12941/36 12941/37 12941/38 12941/39 12941/40 12941/41 12941/42 12941/43 12941/44 12941/45 12941/46 12941/47 12941/48
ELTN12941/36EA 278 PLN
ELTN12941/36 ELTN12941/37 ELTN12941/38 ELTN12941/39 ELTN12941/40 ELTN12941/41 ELTN12941/42 ELTN12941/43 ELTN12941/44 ELTN12941/45 ELTN12941/46 ELTN12941/47 ELTN12941/48
Obuwie ochronne ESD, sznurowane, Basic, jo_Colour Orange Low, Jori
Obuwie i buty
Safety shoes made from hydrophobised, quick drying, tear resistant textile upper combined with reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU (New Basic) sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/orange
Certyfikaty: EN 61340 (ESD); EN ISO 20345; S3, SRC.