tiocyjanian amonu ≥97.5% ACS
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.
Purity: 97.5% min
CAS No.: 1762-95-4
Molecular Formula: NH4SCN
Formula Weight: 76.12
Physical Form: Crystalline
Density: 1.305
Melting Point: 147-149°
Solubility: Soluble in water, ethanol, methanol, ammonia, and acetone. Practically insoluble in CHCl3 and ethyl acetate.
Beilstein Reference: 3595135
Merck Reference: 14,561
MDL No.: MFCD00011428
Fertilizers, photography, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe
Solubility: Soluble in water, ethanol, methanol, ammonia, and acetone. Practically insoluble in CHCl3 and ethyl acetate.
Formuła:
NH₄SCN Temp. topnienia: 147…149 °C Gęstość: 1,305 Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00011428 Numer CAS: 1762-95-4 Numer WE (dawny EINECS): 217-175-6 Klasyfikacja Merck : 14,561 |