N,N-Dietyloacetamid ≥99%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
Synonim:
N-Acetyldiethylamine
B20762.14
B20762.22
B20762.36
B20762.14EA
292
PLN
B20762.14
B20762.22
B20762.36
N,N-Dietyloacetamid ≥99%
N,N-Dietyloacetamid
Purity: 99%
CAS No.: 685-91-6
Molecular Formula: CH3CON(CH2CH3)2
Formula Weight: 115.18
Flash Point: 76°(169°F)
Refractive Index: 1.4400
Boiling Point: 182-186°
Beilstein Reference: 1209428
MDL No.: MFCD00009047
Formuła:
C₆H₁₃NO Temp. Wrzenia: 182…186 °C Gęstość: 0,92 Temp. Zapłonu: 76 °C (169 °F) Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00009047 Numer CAS: 685-91-6 Numer WE (dawny EINECS): 211-685-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...