Drukuj
Pipetting system for 96- and 384-well plates, PLATEMASTER®
Dostawca: GILSON
F110761 F110762
631-1375EA 80320 PLN
631-1375 613-5913
Pipetting system for 96- and 384-well plates, PLATEMASTER®
Automated Liquid Handling Systems
This compact, ergonomic system is an easy to use and accurate solution for high throughput pipetting of 96- and 384-well plates.
  • Helps to save valuable time, money and resources in the lab
  • Filling a 96-well plate requires less than 10 seconds
  • Fill a 384-well plate in less than a minute, in only 4 pipetting steps
  • Greatly reduces the number of pipetting steps necessary to fill a microplate, when compared to using regular multi-channel pipettes
  • Doesn't use a motor or require electricity, so it performs high-throughput pipetting silently
  • Reduce risk of human errors by eliminating the possibility of skipping or repeating wells
  • All enzymatic reactions or cell-based applications can be started and stopped in the 96 wells at the same time
By using high quality PIPETMAN® tips, an accuracy and precision level equaling an 8- or 12-channel pipette is achieved in a single motion. Additionally, the risk of double-pipetting entire rows or columns is avoided.
Through a variety of accessories such as adapters for loading 384-well microplates, reagent reservoirs or sample heater blocks, the PLATMASTER® can be fully adapted to a variety of application needs.
Recommended working temperature: 20 to 25 °C
Dimensions (W×L×H): 300×450×400 mm
Weight: 16 kg