Drukuj
Fiolki do autosamplera, krótki gwint, plastikowe, TopSert™, Supelco®
Dostawca: Merck
SU860160 SU860181 SU860182 SU861132
SIALSU860160EA 0 PLN
SIALSU860160 SIALSU860181 SIALSU860182 SIALSU861132
Fiolki do autosamplera, krótki gwint, plastikowe, TopSert™, Supelco®
Vialki
These vials are made of PP with graduations and 9 mm fitting.
  • Clear or amber
  • Large opening of 6 mm