Drukuj
Pistolety natryskowe
Dostawca: ENTEGRIS
421-22-11 421-22-21 421-32-21 421-32-31 421-32-11
ENTE421-22-11EA 0 PLN
ENTE421-22-11 ENTE421-22-21 ENTE421-32-21 ENTE421-32-31 ENTE421-32-11
Pistolety natryskowe
Pistolety natryskowe
Made of high purity PFA and PTFE (all wetted parts).
  • With 4,8 mm orifice and 6,35 and 12,7 mm FNPT inlet connections
  • Max. pressure 5,52 bar
  • Coloured caps enable visual chemical differentiation at a glance