Drukuj

Anti-TNFRSF13C Mouse Monoclonal Antibody [clone: HuBR9.1]

Dostawca: ProSci
PRSI36-115EA 2930 PLN
PRSI36-115
Anti-TNFRSF13C Mouse Monoclonal Antibody [clone: HuBR9.1]
Przeciwciała
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR