Drukuj
Worki osuszające z żelem krzemionkowym, Minipax®
Dostawca: STROEBEL GMBH
669039AG37 669039AG41 669039AG33 669039AG43 669039AG53 669039AG49
STRO669039AG37EA 1620 PLN
STRO669039AG37 STRO669039AG41 STRO669039AG33 STRO669039AG43 STRO669039AG53 STRO669039AG49
Worki osuszające z żelem krzemionkowym, Minipax®
Eksykatory Wkłady osuszające
These robust, compact and versatile desiccant bags are supplied with silica gel used for pharmaceutical products and test tubes, as well as for clinical diagnoses and electrical tools.
Applications: Electronics industry, pharmaceutics and diagnostics, precision optics, aviation and defense technology.
Minipax® is probably the best known desiccant solution on the market. They are produced from high quality and high performance silica gel and are approved desiccants by the US FDA. Therefore they are recommended for the use in pharmaceuticals, vitamin and food products. At room temperature (18 to 35 °C) and high relative humidity (60 to 100%), silica gel can adsorb about 35% of its own weight in humidity.
In combination with composite aluminium foil, they can reduce the relative humidity inside to below 40%. Minipax® are environmentally friendly, non-toxic and can therefore be disposed of with normal waste.