Drukuj
Akcesoria do systemów atomizacji płomieniowej
Dostawca: VARIAN
9910025700 9910093000 9910093100 9910093400 9910093500 9910093600 9910105800 6910009700
VARI9910025700EA 1110 PLN
VARI9910025700 VARI9910093000 VARI9910093100 VARI9910093400 VARI9910093500 VARI9910093600 VARI9910105800 VARI6910009700
Akcesoria do systemów atomizacji płomieniowej
Spektrofotometry i fotometry Fotometry płomieniowe
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Informacje do zamówienia: Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.