Drukuj
Detergent do ręcznego czyszczenia, płynny, neodisher® LM 3
Dostawca: Dr. Weigert
440030 440049
148-3162EA 1200 PLN
148-3162 141-0940
Detergent do ręcznego czyszczenia, płynny, neodisher® LM 3
Środki czystości Detergenty i środki myjące
For manually cleaning and pre-cleaning of laboratory glassware as well as for the cleaning of pipettes and viscometers in special washers. Especially suitable for the cleaning of laboratory glassware in water, phosphates and metal laboratories and laboratories where enzymatic and serological analyses are conducted.
  • Phosphate-free
  • Components: <5% amphoteric surfactants
  • Dosage: 0,2 to 20%