Drukuj

Anti-IgY Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dostawca: Tonbo Biosciences
35-8081-M002EA 732 PLN
35-8081-M002
Anti-IgY Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Goat anti-Chicken IgY (H&L) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Chicken
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR