Drukuj

Anti-IgG Goat Polyclonal Antibody (Biotin)

Dostawca: Tonbo Biosciences
30-8055-U500EA 892 PLN
30-8055-U500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Przeciwciała
Goat anti-Mouse IgG (H&L), F(ab')2 Fragment - Affinity Pure, Biotin Conjugate, min x w/human IgG or serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR