Drukuj
Płyty grzewcze, seria C-MAG HP
Dostawca: IKA
3581800 3582002 3581600 3581602 3581603 3581802 3581803 3582000 3582003
453-0087EA 4020 PLN
453-0087 553-0002 453-0070 553-0000 453-0071 553-0001 453-0072 453-0073 453-0074
Płyty grzewcze, seria C-MAG HP
Urządzenia grzewcze Płyty grzejne
Compact hotplates with a chemically resistant ceramic top. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature a digital error code display and 'hot' top indicator to warn user that the surface is hot.
  • C-MAG HP 7 and C-MAG HP 10 can be used with e.g. ETS-D5 contact thermometer, enabling precise temperature control
  • Fixed safety circuit shuts off heater if temperature rises above 550 °C
  • Microprocessor control sends feedback to the hotplate to maintain set temperature
  • Digital display ensures accurate and repeatable temperature settings
IP protection class according to DIN EN 60529: IP 21
Certyfikaty: IP protection class according to DIN EN 60529: IP 21