Drukuj
Filtr do X-plore® 3300/3500 i X-plore® 5500
Dostawca: Dräger
6738001 6738002 6738011 6738353 6738354 6738391 6738875
111-0428EA 172 PLN
111-0428 111-0427 111-0423 111-0422 111-0421 111-0418 111-0911
Filtr do X-plore® 3300/3500 i X-plore® 5500
Maski oddechowe Maski wielokrotnego użytku Filtry do masek
The X-plore® twin filter series with bayonet connections successfully combines comfort with practicality. The filters are suitable for Dräger X-plore® 3300/3500 half masks and X-plore® 5500 full face masks.
  • White particle filters for easy identification
  • Excellent user safety as they can also be worn under visors
  • 'R' classification: Permitted for reuse (more than one layer)
  • Robust filter with blue ABS plastic housing and safe bayonet connection with clear guidance to bring the filter securely into place
EN 14387/2000/A1:2006
Certyfikaty: EN 14387/2000/A1:2006