Drukuj
Kolumny HPLC, Poroshell 120 EC-C8
Dostawca: Agilent
699775-906 691775-906 691975-306 691975-906 693775-906 693975-306 693975-906 695775-906 695975-306 695975-906 697775-906 697975-306 697975-906 699975-306 699975-906 823750-913 821725-913 820750-913 699970-306 699970-906 695770-906 695970-306 695970-906 699770-906
HEWL699775-906EA 4510 PLN
HEWL699775-906 HEWL691775-906 HEWL691975-306 HEWL691975-906 HEWL693775-906 HEWL693975-306 HEWL693975-906 HEWL695775-906 HEWL695975-306 HEWL695975-906 HEWL697775-906 HEWL697975-306 HEWL697975-906 HEWL699975-306 HEWL699975-906 HEWL823750-913 HEWL821725-913 HEWL820750-913 HEWL699970-306 HEWL699970-906 HEWL695770-906 HEWL695970-306 HEWL695970-906 HEWL699770-906
Kolumny HPLC, Poroshell 120 EC-C8
Kolumny chromatograficzne
Poroshell 120 is made with a superficially porous particle that has a solid silica core and a porous outer layer. This particle provides high efficiency at lower pressures and is ideal for fast or high-resolution separations of many types of analytes.
  • Less retentive for non-polar samples
  • Exhaustive endcapping ideal for use with basic compounds, for those producing poor peak shapes on other columns
  • Ideal for a wide range of applications
The EC-C8 bonded phase is made by first chemically bonding a dense monolayer of dimethyl-n-octyl silane stationary phase to the porous shell of the Poroshell 120 silica support. The bonded phase packing is then endcapped using proprietary reagents and procedures to obtain maximum deactivation of the silica surface.