Drukuj
Gazoszczelne strzykawki, od 10 do 500 µl
Dostawca: Trajan Scientific and Medical
002200 002208 002250 002252 002455 002500 003200 003250 004200 004229 004230 004250 004279 005200 005229 005230 005250 005279 006200 006229 006230 006250 006279 007200 007229 007230 007250 002202 004232 005232 006232 007232 002200CAL 003200CAL 004200CAL 006200CAL 005200CAL 007200CAL
549-0288EA 321 PLN
549-0288 549-0445 549-0289 562-1031 549-0448 549-0449 549-0295 549-0296 549-0300 549-0301 549-0302 549-0303 549-0304 549-0310 549-0311 548-0940 549-0312 549-0313 549-0318 549-0319 549-0320 549-0446 549-0321 549-0326 548-0945 548-0946 549-0447 548-0935 548-0938 548-0941 548-0942 548-0947 SGEA002200CAL SGEA003200CAL SGEA004200CAL SGEA006200CAL SGEA005200CAL SGEA007200CAL
Gazoszczelne strzykawki, od 10 do 500 µl
Strzykawki i igły Strzykawki do chromatografii
Constructed from borosilicate glass, PTFE plunger tips and Kel-F® or PTFE Luer cones where applicable.
  • Assembled with an interference fit between plunger tip and barrel to effect the gas sealing properties of the syringe
  • Plungers are replaceable on these syringes (plungers on request)
Barrel OD: 10 µl = 6,5 mm; 25 to 500 µl = 8 mm