Drukuj
Wielokanałowe pipety korekcyjne z regulowanym rozstawem końcówek, elektroniczne, z bluetooth, E1-ClipTip™
Dostawca: Thermo Fisher Scientific
4672010BT 4672020BT 4672030BT 4672040BT 4672050BT 4672060BT 4672070BT 4672080BT 4672090BT 4672100BT
613-5767EA 10000 PLN
613-5767 613-5770 613-5768 613-5771 613-5764 613-5769 613-5772 613-5765 613-5763 613-5766
Wielokanałowe pipety korekcyjne z regulowanym rozstawem końcówek, elektroniczne, z bluetooth, E1-ClipTip™
Pipety Pipety i dozowniki seryjne
These pipettes are ideal for quickly and efficiently performing sample transfers between virtually any tube, rack, microplate or horizontal gel box. Adjustable tip spacing allows the user to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • ClipTip Interlocking Technology: Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every application: Step based Matrix or Presets with settings for most common applications
  • Five user profiles, personalised user interface and password protection offer unified operation for multiple users and environments
  • Ability to store up to 30 common protocols with a specific name and calibration in the Programs function
  • Service and calibration trackers remind users when the pipettor requires service or repair
  • Lithium-ion battery ensures long-lasting performance; unit may be recharged by a charging stand or a plug
  • Pipettes can be programmed via PC by connecting to the new My Pipette™ Creator app via Bluetooth* or USB connection
Informacje do zamówienia: Supplied with universal power supply. Two-year extended warranty only available via web registration.
Uwaga: Please note: ClipTip™ 384 and ClipTip™ 12,5 µl tips and compatible E1-ClipTip™ models do not have interlocking technology.
* Bluetooth models are not available in all countries.