Drukuj
Buty ochronne ESD do kostki, sznurowane, E-Sport, JALAS® 1625
Dostawca: EJENDALS
1625-39 1625-42 1625-43 1625-44 1625-47 1625-36 1625-37 1625-38 1625-40 1625-41 1625-45 1625-46
EJEN1625-39EA 0 PLN
EJEN1625-39 EJEN1625-42 EJEN1625-43 EJEN1625-44 EJEN1625-47 EJEN1625-36 EJEN1625-37 EJEN1625-38 EJEN1625-40 EJEN1625-41 EJEN1625-45 EJEN1625-46
Buty ochronne ESD do kostki, sznurowane, E-Sport, JALAS® 1625
Obuwie i buty
Anti-static, lightweight, ESD safety ankle boots made from PU coated split leather with EVA, polyester-based conductive thread insole, polyester/polyamide lining plus a PU outsole.
  • Orthopaedic insoles possible
  • Ergothan shock absorption zone
  • Toe protection cap made from aluminium
  • Flexible, puncture resistant insole made from plasma-treated composite (PTC)
Mainly used for outdoors, indoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Certyfikaty: EN 61340-5-1 (ESD); BGR191; EN ISO 20345:2011; S3, SRC, CI