Drukuj
Sandały ochronne ESD, zapięcie na rzepy, białe, JALAS® 3500
Dostawca: EJENDALS
3500-36 3500-37 3500-38 3500-39 3500-40 3500-41 3500-42 3500-43 3500-44 3500-45 3500-46 3500-47
EJEN3500-36EA 0 PLN
EJEN3500-36 EJEN3500-37 EJEN3500-38 EJEN3500-39 EJEN3500-40 EJEN3500-41 EJEN3500-42 EJEN3500-43 EJEN3500-44 EJEN3500-45 EJEN3500-46 EJEN3500-47
Sandały ochronne ESD, zapięcie na rzepy, białe, JALAS® 3500
Obuwie i buty
Anti-static, lightweight, ESD safety sandals made from textile PU coated leather with EVA, polyester-based conductive thread insole and polyester/polyamide lining.
  • Double shock-absorption zones in Poron® XRD®
  • Plastic shank insole and PU outsole
  • Polstered shaft edge and ventilating insole
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for laboratory and paint spraying work.
Colour: White/black
Certyfikaty: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC