Drukuj
Mikropłytki 384-dołkowe dla immunologii, BRANDplates®
Dostawca: Brand
781740 781741 781742
735-2051EA 1560 PLN
735-2051 735-2052 735-2053
Mikropłytki 384-dołkowe dla immunologii, BRANDplates®
Mikropłytki Mikropłytki i paski immunologiczne
PS, solid or with transparent base. These flat bottom plates are available in different colours, and with standard or transparent base formats.
immunoGrade™: The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
Strongly hydrophilic surface. Especially well suited for binding molecules with predominantly hydrophilic regions. Plates with a hydroGrade™ surface can be used in liquid phase assays with hydrophobic components.