Drukuj
1536-dołkowe mikropłytki UHTS, niepoddane obróbce, BRANDplates®
Dostawca: Brand
781640 781641 781642
735-2019EA 4680 PLN
735-2019 735-2020 735-2021
1536-dołkowe mikropłytki UHTS, niepoddane obróbce, BRANDplates®
Mikropłytki Mikropłytki do wysokosprawnych testów przesiewowych
These PS flat bottom plates are available sterile or non sterile.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analyses
  • Sterile plates are supplied individually wrapped with lid
pureGrade™: Non treated, medium binding, non sterile surface. These plates are suitable for homogeneous assays, screening and storage.
Non treated, medium binding, non sterile surface. These plates are suitable for homogeneous assays, screening and storage.
Non treated, medium binding, sterile surface. Sterilised by β radiation, these plates are especially well suited for bacteriological assays.
Certyfikaty: BRANDplates® are non cytotoxic according to ISO 10993-5, free of endotoxins (<0.01 EU/ml), and free from DNA, DNase and RNase. Sterile products comply with ISO 11137 and AAMI guidelines, SAL 10⁻⁶.