Drukuj
96-dołkowe mikropłytki do hodowli komórkowych, BRANDplates®
Dostawca: Brand
781902 781910 781911 781960 781961 781962 781965 781968 781970 781971 782022 782030 782031 782082 782090 782091 781900 781912 781913 781975 782095 782094 782034 782035 781974
735-2083EA 2760 PLN
735-2083 735-2090 735-2091 735-2097 735-2098 735-2099 735-2101 735-2102 735-2103 735-2104 735-2116 735-2119 735-2120 735-2124 735-2127 735-2128 735-2082 733-2404 733-2405 733-2399 733-2403 733-2402 733-2400 733-2401 733-2398
96-dołkowe mikropłytki do hodowli komórkowych, BRANDplates®
Mikropłytki Płytki do hodowli komórkowych
PS, solid or with transparent base. Plates are available with different well shapes (round U-bottom, conical V-bottom, flat F-bottom or flat C-bottom with curved edges), in different colours, and with a choice of cell culture treated surfaces.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • Standard 96-well plates have coloured alphanumeric code identifying surface type (cell culture treated orange, immuno treated blue, non-treated grey)
  • White plates provide maximum reflection and