Drukuj
PerfeCTa® qPCR FastMix™ mieszanina reakcyjna
Dostawca: Quantabio
95076-012 95076-05K 95078-012 95078-05K 95077-012 95077-05K
733-1394EA 6780 PLN
733-1394 733-1395 733-1402 733-1403 733-1398 733-1399
PerfeCTa® qPCR FastMix™ mieszanina reakcyjna
Odczynniki do kwasów nukleinowych Enzymy i zestawy do qPCR, RT-qPCR
FastMixes include buffers and stabilisers optimised for maximum PCR efficiency, reproducibility, and sensitivity with TaqMan® and TaqMan® MGB probe chemistries. A key component of PerfeCTa® qPCR FastMixes is AccuFast™ Taq DNA polymerase that enables instant activation and rapid polymerisation kinetics. Upon heat activation (20 seconds at 95 °C), the antibodies irreversibly denature releasing fully active, unmodified Taq DNA polymerase. This promotes specific and efficient primer extension with the convenience of room temperature reaction assembly. FastMixes deliver maximum PCR efficiency, sensitivity, specificity, and a robust fluorescent signal with either fast or conventional cycling conditions.