Drukuj
96-dołkowe mikropłytki do immunologii, BRANDplates®
Dostawca: Brand
781720 781721 781722 781724 781731 781780 781782 781840 781842 781732
735-2042EA 953 PLN
735-2042 735-2043 735-2044 735-2046 735-2050 735-2054 735-2055 735-2064 735-2066 738-0191
96-dołkowe mikropłytki do immunologii, BRANDplates®
Mikropłytki Mikropłytki i paski immunologiczne
PS, solid or with transparent base. Plates are available with different well shapes (round U-bottom, conical V-bottom, flat F-bottom or C-flat bottom with curved edges), in different colours, and with a choice of treated surfaces.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • Standard 96-well plates have coloured alphanumeric code identifying surface type (cell culture treated orange, immuno treated blue, non-treated grey)
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analyses
immunoGrade™: The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
hydroGrade™: Strongly hydrophilic surface. Especially well suited for binding molecules with predominantly hydrophilic regions. Plates with a hydroGrade™ surface can be used in liquid phase assays with hydrophobic components.
lipoGrade™: Strongly hydrophobic (lipophilic) surface. Especially suitable for binding molecules with hydrophobic regions. Microplates with lipoGrade™ surfaces can be used in liquid phase assays with hydrophilic components.
Strongly hydrophilic surface. Especially well suited for binding molecules with predominantly hydrophilic regions. Plates with a hydroGrade™ surface can be used in liquid phase assays with hydrophobic components.
Certyfikaty: BRANDplates® are non cytotoxic according to ISO 10993-5, free of endotoxins (<0,01 EU/ml), and free from DNA, DNase and RNase.